Learn More
AdipoGen N4-Acetylsulfamethoxazole
SDP
Supplier:  AdipoGen CDXA0291M010
Encompass_Preferred
Chemical. CAS 21312-10-7. Formula C12H13N3O4S. MW 295.3. Synthetic. This is a metabolite of the sulfonamide bacteriostatic antibiotic sulfamethoxazole. Sulfamethoxazole is metabolized via acetylation, catalyzed by liver extramitochondrial N-acetyl transferases. Acetyl-Sulfamethoxazole is excreted in urine. Both compounds can be used as a probe for the molecular percentage enrichement of liver extramitochondrial acetyl-CoA. Acetyl-Sulfamethoxazole can also be used as a reference for sulfamethoxazole impurity and waste determination.
Catalog No. 50-256-1766
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.