Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N4-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-deoxycytidine 99.0+%, TCI America™
SDP

Catalog No. B30871G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30871G 1 g
B30875G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B30871G Supplier TCI America Supplier No. B30871G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 67219-55-0
Molecular Formula C37H35N3O7
Molecular Weight (g/mol) 633.701
MDL Number MFCD00010114
InChI Key MYSNCIZBPUPZMQ-VOTWKOMSSA-N
Synonym Bz-DMT-dC
PubChem CID 2724490
IUPAC Name N-[1-[(2R,4S,5R)-5-[[bis(4-methoxyphenyl)-phenylmethoxy]methyl]-4-hydroxyoxolan-2-yl]-2-oxopyrimidin-4-yl]benzamide
SMILES COC1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=C(C=C3)OC)OCC4C(CC(O4)N5C=CC(=NC5=O)NC(=O)C6=CC=CC=C6)O

Specifications

Melting Point 119°C
Color White
Quantity 1 g
Formula Weight 633.70
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N4-Benzoyl-5′-O-(4,4′-dimethoxytrityl)-2′-deoxycytidine
EINECSNumber (5)-6799
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.