Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N4-Ethyl-N4-(2-methanesulfonamidoethyl)-2-methyl-1,4-phenylenediamine Sesquisulfate Monohydrate 98.0+%, TCI America™
SDP

Catalog No. E065825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E065825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E065825G Supplier TCI America Supplier No. E065825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 24567-76-8
Molecular Formula C24H48N6O16S5
Molecular Weight (g/mol) 836.974
InChI Key NPDFXFLCEDDWEG-UHFFFAOYSA-N
Synonym N-[2-[(4-Amino-3-methylphenyl)ethylamino]ethyl]methanesulfonamide Sesquisulfate
PubChem CID 91578
IUPAC Name N-[2-(4-amino-N-ethyl-3-methylanilino)ethyl]methanesulfonamide;sulfuric acid
SMILES CCN(CCNS(=O)(=O)C)C1=CC(=C(C=C1)N)C.CCN(CCNS(=O)(=O)C)C1=CC(=C(C=C1)N)C.OS(=O)(=O)O.OS(=O)(=O)O.OS(=O)(=O)O

Specifications

Quantity 25 g
Formula Weight 418.49
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N4-Ethyl-N4-(2-methanesulfonamidoethyl)-2-methyl-1,4-phenylenediamine Sesquisulfate Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.