Learn More
Cole-Parmer Implen N50 NanoPhotometer UV/Vis Spectrophotometer with Touch Screen, 110/220 V
Supplier:  Cole-Parmer 8307056
Encompass_Preferred
For those in need of flexibility, this family of UV/visible spectrophotometers offers reliable protein and DNA measurements for mobile and laboratory applications. Requiring only a minute sample size of 0.3 µL and with a limit of detection down to 0.1 ng/µL (ds DNA) or 0.003 mg/µL (BSA), this spectrophotometer series accommodates any application. Simplified optics with two precisely defined pathlengths provide two fixed and sealed anchor points, prohibiting drift and eliminating the need for recalibration.
Equipped with Wi-Fi, USB A/B, HDMI and LAN interface connections for flexible instrument control and data saving options. Front USB flash drive port offers quick and easy data transfer.
Catalog No. NC1717932
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.