Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam N6-Methyladenosine, Modified Adenosine Analog, Solid, 100 mg
Description
- Chemical Name: N6-Methyladenosine (m6A)
- Product Type: Modified adenosine analog
- Molecular Weight: 281.27 g/mol
- Purity: >97%
- Form: Solid
- CAS Number: 1867-73-8
- Composition: 97.13% (2R,3S,4R,5R)-2-(hydroxymethyl)-5-[6-(methylamino)purin-9-yl]oxolane-3,4-diol
- Biological Significance: Abundant modification in mRNA; found in viruses, eukaryotes including mammals, insects, plants, and yeast; also found in tRNA, rRNA, and snRNA
- Storage: Store at -20°C
- Shipping: Ambient - Can Ship with Ice
Specifications
Specifications
| Target Molecule | N6-Methyladenosine |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Quantity | 100 mg |
| For Use With (Application) | Sample Prep and Detection |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?