Learn More
Abcam N6-Methyladenosine, Modified Adenosine Analog, Solid, >97% Purity, 500 mg
Description
- Molecular Weight: 281.27 g/mol
- Purity: >97%
- Description: Modified adenosine analog methylated at the nitrogen-6 position
- Biological context: Abundant modification in mRNA, found within some viruses, and most eukaryotes including mammals, insects, plants and yeast; also found in tRNA, rRNA, and small nuclear RNA (snRNA)
- Composition: 97.13% (2R,3S,4R,5R)-2-(hydroxymethyl)-5-[6-(methylamino)purin-9-yl]oxolane-3,4-diol
- Form: Solid
- CAS Number: 1867-73-8
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Target Molecule | N6-Methyladenosine |
| Quantity | 500 mg |
| For Use With (Application) | Sample prep and detection |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.