Learn More
QA Bio Inc NA2 Glycan, 2-AA labeled
Supplier:  QA Bio Inc CAANA201
Encompass
NA2 Glycan Synonyms: NA2 N-linked oligosaccharide. G2, A2G2, A2G(4)2. • Description: Asialo- bi-antennary complex-type N-glycan (oligosaccharide). • Molecular Weight: 1642 • Purity: >90% pure as assessed by a combination of 1 H-NMR and HPLC. • Sources: NA2 glycan is found on many mammalian glycoproteins including IgG, gamma globulins, and many serum glycoproteins. This product is typically purified from the oligosaccharide pool released from bovine serum by hydrazinolysis using a combination of HPLC and glycosidase digestion. • Form: Dry. Dried by centrifugal evaporation from an aqueous solution.
Catalog No. 50-223-6521
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.