Learn More
Cayman Chemical PurIn rIbosIde5OtrIphosp 2.5mg
Supplier: Cayman Chemical 384912.5 MG
An antimycobacterial agent active against M. tuberculosis and M. bovis (MICs 0.38 and 1.6 UM respectively)
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.