Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nacolomab Tafenatox Recombinant Mouse Monoclonal Antibody
GREENER_CHOICE

Catalog No. PIMA541900
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
PIMA541900 100 μg
1 options
This item is not returnable. View return policy

Catalog No. PIMA541900

Supplier: Invitrogen™ MA541900

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Recombinant Monoclonal Antibody

Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.

Nacolomab Tafenatox is a biosimilar that targets MUC1 sialylated CA242.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nacolomab Tafenatox
Applications ELISA
Classification Recombinant Monoclonal
Concentration 0.63 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias r-c242Fab-SEA
Host Species Mouse
Immunogen Human MUC1 sialylated CA242.
Purification Method Protein L
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 F(ab) κ
Target Molecule Chemical
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.