Learn More
MBL International Corporation CycLex NAD+/NADH Colorimetric Assay Kit Ver.2
SDP
Supplier:  MBL International Corporation CY1253V2
Encompass_Preferred
Size:100 Assays; Target:NAD+/NADH; This Kit provides a convenient method for sensitive measurement of NAD+, NADH and their ratio in cell extracts. This kit is designed for specific detection of NAD+ and NADH by an enzyme cycling reaction.
Catalog No. 50-455-74
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.