Learn More
Medchemexpress LLC NAD P H-flavin reduc 20ug

Supplier: Medchemexpress LLC HYP7170920UG
NAD(P)H-flavin reductase is a recombinant His-tagged enzyme expressed in E. coli that catalyzes the reduction of soluble flavins using NADH or NADPH. Supplied as a lyophilized powder formulated in PBS with trehalose, it is intended for enzymatic assays and biochemical studies requiring a reliable flavin reductase reference reagent.
- Recombinant enzyme with N-terminal His tag.
- Catalyzes reduction of soluble flavins using NADH or NADPH.
- Supplied lyophilized from PBS with 6% trehalose.
- Purity greater than 90% by reducing SDS-PAGE.
- Low endotoxin level (<1 EU/μg).
- Stable when stored at -20°C; aliquot and freeze for long-term storage.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.