Learn More
AdipoGen NADE B subtilisrec
SDP
Supplier:  AdipoGen AG40T0109C050
Encompass_Preferred
Protein. Full-length NAD synthetase Bacillus subtilis. Source E. coli. Endotoxin content N/A. Liquid. In 50mM TRIS-HCl, pH 8, containing 75mM sodium chloride, 5% glycerol and 5mM DTT. Purity >98% SDS-PAGE. NAD synthetase is an essential enzyme involved in both the de novo biosynthesis and salvage of NAD, catalyzing the final step of both pathways. Since NAD has a vital role in cell metabolism, the enzyme represents a valid target for the development of new antimycobacterial agents. It was shown to be induced by heat shock, salt stress, oxidative stress, glucose limitation and oxygen limitation and is involved in biological processes like stress response and sporulation resulting in formation of a cellular spore. Catalytic activity ATP deamido-NAD NH3 = AMP diphosphate NAD
Catalog No. 50-256-2758
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.