Learn More
Enzo Life Sciences NADP. disodium salt (250mg). CAS: 24292-60-2
Supplier: Enzo Life Sciences ALX480003M250
Alternative name: beta-Nicotinamide Adenine Dinucleotide Phosphate, beta-NADP, Coenzyme II. Purity: ≥97% (calc based on dry substance, enzymatic). Appearance: White to light yellow powder. Solubility: Soluble in water (50mg/ml). Long Term Storage: -20°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.