Learn More
Bioassay Systems EnzyFluo™ NADP/NADPH Assay Kit. For quantitative determination of NADP and NADPH and evaluation of drug effects on their metabolism (This product is a replacement for the discontinued product EFNP-100).
SDP
Catalog No. 502255874 Shop All Bioassay Systems Products
Encompass_Preferred
For quantitative determination of NADP and NADPH and evaluation of drug effects on their metabolism (This product is a replacement for the discontinued product EFNP-100). Key Features: Sensitive and accurate. Detection limit of 0.01 µM and linearity up to 1 µM NADP+/NADPH in 96-well plate assay. Convenient. The procedure involves adding a single working reagent, and reading the fluorescence at time zero and 30 min. Room temperature assay. High-throughput. Can be readily automated as a high-throughput 96-well plate assay for thousands of samples per day. Method: FL530/585nm. Samples: Cell, tissue extracts etc. Species: All. Procedure: Assay takes 30 min. Kit size: 100 tests. Detection limit: 0.01 µM.
This item is not returnable. View return policy
Catalog No. 50-225-5874 Supplier Bioassay Systems Supplier No. EF2NP100
For quantitative determination of NADP and NADPH and evaluation of drug effects on their metabolism (This product is a replacement for the discontinued product EFNP-100). Key Features: Sensitive and accurate. Detection limit of 0.01 µM and linearity up to 1 µM NADP+/NADPH in 96-well plate assay. Convenient. The procedure involves adding a single working reagent, and reading the fluorescence at time zero and 30 min. Room temperature assay. High-throughput. Can be readily automated as a high-throughput 96-well plate assay for thousands of samples per day. Method: FL530/585nm. Samples: Cell, tissue extracts etc. Species: All. Procedure: Assay takes 30 min. Kit size: 100 tests. Detection limit: 0.01 µM.