Learn More
Enzo Life Sciences Nafamostat . mesylate (50mg). CAS: 82956-11-4
SDP
Supplier:  Enzo Life Sciences BMLPI1430050
Encompass_Preferred
Competitive, reversible broad-spectrum serine proteinase inhibitor that can be a used as a specific tryptase inhibitor at very low concentrations. Has been used in vitro and in vivo. Purity: ≥98% (HPLC). Solubility: Soluble in water (25 mg/ml) Note: Phosphate-containing buffers may cause precipitation. Long Term Storage: Ambient.
Catalog No. 50-201-1483
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.