Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Autoclavable Graduated Buckets
GSA_VAGREENER_CHOICE

Catalog No. 0368722
Change view
Click to view available options
Capacity (Metric):
7.6 L
10.4 L
13.2 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
03-687-22 7.6 L
03-687-20 10.4 L
03-687-24 13.2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 03-687-22 Supplier Thermo Scientific™ Supplier No. 70120080
Add to Cart
Add to Cart

Contain a variety of substances, including chemical and corrosive liquids, in these graduated buckets that are sturdy and autoclavable.

Contain a variety of substances, including chemical and corrosive liquids, in sturdy and convenient Thermo Scientific™ Nalgene™ Autoclavable Graduated Buckets, which are a lab necessity. These general-purpose, polypropylene laboratory buckets are fully autoclavable and provide excellent chemical resistance.

  • White, polypropylene pails offer sturdy construction for long-lasting use in the lab.
  • Nylon-coated, wire bail handle provides a comfortable and sturdy grip.
  • Molded-in graduated in pints and liters are easy to read.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 7.6 L
Graduations Molded
Material Polypropylene
Autoclavable Autoclavable
Capacity (English) 8 qt.
Description Bucket Graduated PP 7.6 L
Includes Handle
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.