Learn More
Thermo Scientific™ Nalgene™ Closed-Dome Polypropylene Tanks

Description
Store, mix and dispense reagents in Thermo Scientific™ Nalgene™ Closed-Dome Polypropylene Tanks, designed for use as closed containment systems. Offered in sizes ranging from 75 to 380L , tanks feature a 150mm neck opening fitted with a gasketed closure for secure sealing.
Features:
- Tanks may be fitted with spigots
- Autoclavable tanks and closures feature polypropylene construction
Compatible with:
Use with Closed-Dome Tank Closure w/ Mixer Support Assembly (Mfr. No. 2651-0200), BioTech Overhead Mixer (Mfr. No. 2653-0010 and 2653-0020) and Lower Assemblies for BioTech Mixers (Mfr. No. 2654-0030, 2654-0055, 2654-0075 & 2654-0100).
Specifications
Specifications
| Type | Closed-dome Tank |
| Certifications/Compliance | Comply with FDA Reg. 177.1520 and USP Class VI |
| Diameter (Metric) Outer | 72 cm |
| Diameter (English) Outer | 28.5 in. |
| Material | Polypropylene |
| Capacity (English) | 100 gal. |
| Capacity (Metric) | 378.5 L |
| Height (English) | 52 in. |
| Height (Metric) | 132.1 cm |
| No. per Case | 1 |
| Show More |
Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.