Learn More
Thermo Scientific™ Nalgene™ Reusable Analytical Funnels
GSA_VAGREENER_CHOICE
Use this filter holder with funnel with any filtering flask or manifold that accepts a suitable rubber stopper with hole.
$321.50 - $476.50
Specifications
Material PSU
Autoclavable Autoclavable
View More Specs
Products 2
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
09-745
GREENER_CHOICE
View Documents
Thermo Scientific™
DS03104000
250 mL
Each for $321.50
Only null left
Add to Cart
09-746
GSA_VAGREENER_CHOICE
View Documents
Thermo Scientific™
DS03104050
500 mL
Each for $476.50
Only null left
Add to Cart
Description
Description
For use with any filtering flask or vacuum manifold that accepts a rubber stopper with hole. Comes with easy-to-use, no twist vacuum gasket; sterilization and analytical support plates. Accepts 47 ot 50 mm diameter membranes. 1 unit per case.
Features:
  • Reusable, autoclavable polysulfone lasts for years
  • Graduated upper chamber for easy sample measurement
  • Includes analytical and sterilization support plates
  • Use with filtering flasks or manifolds
Includes:
  • Rugged polysulfone funnel
  • Polypropylene copolymer cover with three ports
  • Three friction-fit TPE caps
  • Support plate for 47mm membrane
  • No-twist vacuum silicone gasket which can replace a No. 7 or 8 rubber stopper with 0.56″ (15mm) hole
Specifications
Specifications
PSU
Autoclavable
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.