Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Locator™ Plus Racks for 81-Ampule Cryogenic Box

Catalog No. 1167667
Change view
Click to view available options
Holds:
Cryogenic Box, 81-Ampule
For Use With (Equipment):
For Locator Jr. Plus
For: Locator 4 Plus
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds For Use With (Equipment)
11-676-67 Cryogenic Box, 81-Ampule For Locator Jr. Plus
11-676-68 Cryogenic Box, 81-Ampule For: Locator 4 Plus
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-676-67 Supplier Thermo Scientific™ Supplier No. HR509X19A70
Add to Cart
Add to Cart

Take advantage of efficient, high-volume storage with Thermo Scientific™ Nalgene™ Locator™ Plus Racks for 81-Ampule Cryogenic Box.

Take advantage of efficient, high-volume storage with Thermo Scientific™ Nalgene™ Locator™ Plus Racks for 81-Ampule Cryogenic Box.

  • Designed for long-term storage
  • Features an indexed frame and box organizational system
  • Unique hanging rack promotes easy sample retrieval
  • Suited for both manual and computerized inventory record-keeping methods
TRUSTED_SUSTAINABILITY

Specifications

Color Silver
Length (English) 5.25 in.
Width (English) 5.25 in.
For Use With (Equipment) For Locator Jr. Plus
Holds Cryogenic Box, 81-Ampule
Design Locator Jr. Plus
Type Locator™ Plus Cryogenic Rack
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.