Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Magnetic PVDF Carboy Stirrer
GSA_VA

Catalog No. 14511220 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-511-220 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-511-220 Supplier Thermo Scientific™ Supplier No. DS22270020

missing translation for 'validMainframeMsg'

For Use With: 10 or 20L Nalgene carboys with 83B closure

Mix high volumes of media and buffer solution at low speed using the Thermo Scientific™ Nalgene™ Magnetic PVDF Carboy Stirrer. Adjusts easily to fit either a 10 or 20L Nalgene carboy with an 83B size closure. Supplied with a closure and two impellers. Eliminates the need to retrieve magnets from your solution.

Features:
  • For use with a magnetic stir plate (not included)
  • Includes closure and two impellers
  • Autoclavable

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Closure Size 83B
Description Stir bar D x L: 1/2 x 3 in.
For Use With (Equipment) 10 or 20L Nalgene carboys with 83B closure
Length (English) 3 in.
Length (Metric) 588 mm
Material PTFE, PVDF, Stainless Steel
No. per Case 1 Ea.
No. per Pack 1 Ea.
Product Type Stirrer
Quantity Each

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.