Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Narrow-Mouth HDPE IP2 Bottles with Closure: Bulk Pack
GREENER_CHOICE

Catalog No. 0331332E
Change view
Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
250 mL
500 mL
1000 mL
Closure Size:
20-415 mm
24-415 mm
28-415 mm
38-430 mm
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Size
03-313-32E 500 mL 28-415 mm
03-313-162 30 mL 20-415 mm
14-178-181 60 mL 20-415 mm
03-313-32C 125 mL 24-415 mm
03-313-32D 250 mL 24-415 mm
03-313-32F 1000 mL 38-430 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 03-313-32E Supplier Thermo Scientific™ Supplier No. 3120990016
Add to Cart
Add to Cart

Ship and store liquid reagents using these high density, durable polyethylene bottles with closure. Recommended for customers who design, assemble and certify their own combination packaging.

Ship and store liquid reagents using durable Thermo Scientific™ Nalgene™ Narrow-Mouth HDPE IP2 Bottles with Closure. These high density polyethylene bottles are a safe, economical solution for customers who design, assemble and certify their own combination packaging.

Durable bottles for shipping liquid reagents

  • HDPE bottle with PP closure, bulk pack
  • Narrow mouth
  • Complies with IATA and ICAO regulations

Ordering Information:

Bottles and closures are bulk packed in separate polybags within the same carton

Recommended for:

For storage and shipment of liquid reagents.

TRUSTED_SUSTAINABILITY

Specifications

Capacity 16 oz., 500 mL
Capacity (Metric) 500 mL
Capacity (English) 16 oz.
Color Translucent
Description IP2 Narrow-Mouth Bottles
Diameter (Metric) Inner Neck 21 mm
Diameter (Metric) Inner 21 mm
For Use With (Application) Storage and Shipment of Liquid Reagents
Height (Metric) With Closure 171 mm
Material HDPE
No. per Case 125
No. per Pack 125
Certifications/Compliance Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. Bottles are evaluated at 15psi (103kPa) per 49 CFR 173.27 (c)(2), ICAO Technical Instructions Part 4; 1.1.6, and IATA Dangerous Goods Regulations Section 5.0.2.9.
Closure Included Yes
Mouth Narrow
Shape Round
Height (Metric) 171 mm
Product Type IP2 Bottle
Height (English) 6.7 in.
Closure Material Polypropylene
Closure Size 28-415 mm
Diameter (Metric) 73 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.