Learn More
Thermo Scientific™ Nalgene™ Narrow-Mouth Opaque Amber HDPE Packaging Bottles with Closure: Bulk Pack



Description
Store and ship light-sensitive reagents with Thermo Scientific™ Nalgene™ Narrow-Mouth Opaque Amber HDPE Packaging Bottles with Closure. Ideal for diagnostics, biologicals, specialty chemicals, reagents, adhesives and veterinary pharmaceuticals, these high-density polyethylene bottles are injection blow-molded for uniform wall thickness and strength. They meet USP light transmission standards and are bulk packed for packaging applications.
- Injection blow-molded for uniform wall thickness and strength
- Opaque amber HDPE Bottle with opaque amber PP Closure
- Meets USP light transmission standards
- Tough plastic construction
- Bulk packed for packaging applications
- Leakproof†
Recommended for:
Storage and shipping of light-sensitive reagents.
Ordering Information:
Bottles and closures are packed in separate polybags within the same carton. † The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Capacity | 4 oz., 125 mL |
| Capacity (Metric) | 125 mL |
| Capacity (English) | 4 oz. |
| Color | Opaque Amber |
| Description | Narrow-Mouth Packaging Bottles |
| Diameter (Metric) Inner Neck | 18 mm |
| Material | HDPE |
| No. per Case | 500 |
| No. per Pack | 500 |
| Closure Color | Opaque Amber |
| Show More |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.