Learn More
Thermo Scientific™ Nalgene™ Natural PPCO Diagnostic Bottles without Closure: Bulk Pack

Description
Safely store and handle test kits and many small volume applications with Thermo Scientific™ Nalgene™ Natural PPCO Diagnostic Bottles, for use in sample storage and the shipping of small volume, valuable reagents. These polypropylene copolymer bottles are durable and feature uniform walls resistant to splitting or puncturing. They are packed without closures so they can be matched with various colored closures (Cat. No. 362150 series).
- Nonsterile
- Natural translucent PPCO provides excellent chemical resistance
- Autoclavable
- Leakproof† when assembled with closure (must be ordered separately)
Ordering Information:
Bottles are bulk packed without closures. Packed 2000 per case. Order colored closures separately (Cat. No. 362150).
Specifications
Specifications
| Capacity | 0.5 oz., 15 mL |
| Capacity (Metric) | 15 mL |
| Capacity (English) | 0.5 oz. |
| Color | Natural |
| Diameter (Metric) Inner Neck | 14 mm |
| Material | PPCO |
| No. per Case | 2000 |
| No. per Pack | 2000 |
| Sterility | Non-sterile |
| Neck Finish | 20-415 mm |
| Show More |
Frequently Asked Questions (FAQs)
PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.