Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Oak Ridge High-Speed Centrifuge Tubes made with Teflon™ fluoropolymer and Closure made with Tefzel™
GSA_VAGREENER_CHOICE

Catalog No. 0556216b
Change view
Click to view available options
Capacity (Metric):
10 mL
30 mL
50 mL
Diameter (Metric):
16 mm
25.7 mm
28.8 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Diameter (Metric)
05-562-16B 50 mL 28.8 mm
05-562-16C 10 mL 16 mm
05-562-16A 30 mL 25.7 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Note: Must be filled to 100% of total capacity for centrifugation
Catalog No. 05-562-16B Supplier Thermo Scientific™ Supplier No. 31140050
Only null left
Add to Cart
Add to Cart
Note: Must be filled to 100% of total capacity for centrifugation

Conduct high-speed centrifugation of aggressive chemicals with these tubes, suited for chloroform and phenol extractions in refrigerated machines up to 50,000 x g.

Conduct high-speed centrifugation of aggressive chemicals with Thermo Scientific™ Nalgene™ Oak Ridge High-Speed Centrifuge Tubes made with Teflon™ fluoropolymer and Closure made with Tefzel™, specifically suited for chloroform and phenol extractions.

Features:

  • Teflon™ FEP, Tefzel™ ETFE screw closure
  • Withstand temperatures from -100° to +150°C (-148° to +302°F)
  • Leakproof† Tefzel ETFE closures prevent material loss under ordinary use for most spins <10,000 × g
  • Sealing caps available for 30mL and 50mL size to assure leakproof service especially at high speeds >10,000 × g or when spinning hazardous materials
  • Autoclavable/biohazard/leakproof†/PTFE FEP

Ordering Information:

Optional sealing caps available: DS3131-0020 (for 30mL tube) and DS3131-0024 (for 50mL tube). Cat. No. 3114-0050 has a brim capacity of only 42mL but is designed to fit standard rotor.

Includes:

Tefzel ETFE screw caps. Additional sealing cap assembly available separately (DS3131-series).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Closure Material ETFE
Capacity (Metric) 50 mL
Material FEP
No. per Case 10
No. per Pack 2
Sterility Non-sterile
Autoclavable Autoclavable
Shape Round
Bottom Shape Round
Closure Type Screw Cap
Writing Area No
Diameter (Metric) 28.8 mm
Graduated No
Product Type Centrifuge Tube
Max. RCF 50,000 x g (with sealing cap assembly)
Show More Show Less

Teflon™ is a trademark of The Chemours Company FC, LLC used under license by Nalge Nunc International Corporation.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes.

Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.