Learn More
Thermo Scientific™ Nalgene™ Oak Ridge White Polycarbonate Conical Centrifuge Tube Adapter
GSA_VAGREENER_CHOICE
Catalog No. 0100313 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (Metric):
35 mL
Catalog No. Capacity (Metric)
01-003-13 35 mL
1 options
Catalog No. 01-003-13 Supplier Thermo Scientific™ Supplier No. DS31470050
  • $125.70
    $73.65 / Case of 6
    Save $52.05
    41% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Easily remove the Thermo ScientificTM NalgeneTM Oak Ridge White Polycarbonate Conical Centrifuge Tube Adapter from rotors after use.
Easily remove the Thermo Scientific™ Nalgene™ Oak Ridge White Polycarbonate Conical Centrifuge Tube Adapter from rotors after use. This autoclavable adapter must be used with Nalgene 35L conical Oak Ridge Tubes (Cat. Nos. 3146, 3148).
Ordering Information:
To assure leakproof service, especially at speeds greater than 10,000 x g or when spinning hazardous materials, use Nalgene sealing cap assemblies DS3131-0020 or -0024.
Includes:
Supplied with linerless, polypropylene screw closure which will not leak during ordinary use.
Requires:
Both tubes require white polycarbonate tube adapter (see DS3147 0050 below) to centrifuge.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 35 mL
Material Polycarbonate
No. per Case 6
No. per Pack 6
Autoclavable Autoclavable
Shape Conical
Bottom Shape Conical
Product Type Centrifuge Tube Adapter
Max. RCF 50,000 x g