Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Opaque Amber HDPE Diagnostic Bottles without Closure: Bulk Pack
GREENER_CHOICE

Catalog No. 0331385A
Change view
Click to view available options
Capacity (Metric):
4 mL
8 mL
15 mL
Neck Finish:
13-415 mm
20-415 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Neck Finish
03-313-85A 8 mL 20-415 mm
03-313-85C 4 mL 13-415 mm
03-313-85B 15 mL 20-415 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 03-313-85A Supplier Thermo Scientific™ Supplier No. 3620049025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Safely store and ship light-sensitive, small-volume, diagnostic samples and valuable reagents with these opaque, amber HDPE bottles.

Safely store and ship light-sensitive, diagnostic samples and valuable reagents in small volumes with Thermo Scientific™ Nalgene™ Opaque Amber HDPE Diagnostic Bottles, used in test kits and many small-volume applications. These opaque, amber, high-density polyethylene bottles are for use with light-sensitive reagents and are packed without closures so they can be matched with various colored closures (Cat. No. 362150 series).

  • Nonsterile
  • Durable, uniform walls resistant to splitting or puncturing
  • Leakproof† when assembled with closure (must be ordered separately)

Ordering Information:

Bottles are bulk packed without closures. Packed 2,000 per case. Order colored closures separately (Cat. No. 362150).

TRUSTED_SUSTAINABILITY

Specifications

Capacity 0.25 oz., 8 mL
Capacity (Metric) 8 mL
Capacity (English) 0.25 oz.
Color Opaque Amber
Description 8mL, 20-415 neck finish, bulk pack
Diameter (Metric) Inner Neck 14 mm
Material HDPE
No. per Case 2000
No. per Pack 2000
Sterility Non-sterile
Neck Finish 20-415 mm
Certifications/Compliance Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Classs VI. Product meets the requirements for light-resistant containers per USP latest edition.
Closure Included No
Graduated No
Mouth Narrow
Shape Round
Height (Metric) 45 mm
Product Type Sample Container
Width (English) 0.98 in.
Height (English) 1.77 in.
Closure Material Polypropylene
Closure Size 20-415 mm
Diameter (Metric) 25 mm
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.