Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ PETG Serum Vials with Continuous Thread: Nonsterile, Shrink-Wrapped Modules
GREENER_CHOICE

Catalog No. 03317
Change view
Click to view available options
Volume (Metric):
5 mL
10 mL
20 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
03-317 10 mL
03-316 5 mL
14-178-176 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 03-317 Supplier Thermo Scientific™ Supplier No. 3220320010
Add to Cart
Add to Cart

Protect against leakage, breakage and contamination by using PETG nonsterile serum vials with continuous thread closures.

Protect against leakage, breakage, and contamination using Thermo Scientific™ Nalgene Nonsterile Serum Vials. Lighter than glass. Use with lined continuous thread closures (available separately).

Features:

  • PETG
  • Noncytotoxic
  • Nonpyrogenic
  • Packaged in trayless shrink-wrap modules
  • USP VI-compliant
  • Nonsterile
  • Leakproof† with Nalgene lined continuous-thread closures (available separately)

Ordering Information:

Serum vials are packed in trayless shrink-wrapped modules. Order closures separately (Cat. No. 312158).

TRUSTED_SUSTAINABILITY

Specifications

Description Serum Vials
Diameter (Metric) Inner 11 mm
Color Clear
Diameter (Metric) Outer 24 mm
Material PETG
Sterility Non-sterile
Volume (Metric) 10 mL
No. per Case 1260
No. per Pack 252
Volume (English) 1/3 oz.
Product Line Nalgene
Volume (Metric) Total 10 mL
Closure Type Screw Cap (Not Included)
Quantity 252 Pk., 1260 Cases
Closure Size 20-415
For Use With (Equipment) Lined Continuous Thread Closure
Certifications/Compliance ISO 13485:2003
Height (Metric) 54 mm
Product Type Serum Vial
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.