Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ PMP Erlenmeyer Flasks with Closure
GREENER_CHOICE

Catalog No. 100425G
Change view
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
1000 mL
Closure Size:
33 mm
38 mm
43 mm
53 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Size
10-042-5G 500 mL 43 mm
10-042-5C 125 mL 33 mm
10-042-5E 250 mL 38 mm
10-042-5J 1000 mL 53 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 10-042-5G Supplier Thermo Scientific™ Supplier No. 41090500
Add to Cart
Add to Cart

Perform titrations, preparation and storage of culture media with these flasks, which provide a combination of chemical resistance, strength and transparency.

Perform titrations, preparation and storage of culture media with Thermo Scientific™ Nalgene™ PMP Erlenmeyer Flasks with Closure, which provide a combination of chemical resistance, strength and transparency. Rugged, shatter-proof construction combined with light weight make these flasks a valuble element of lab safety programs.

Features:
  • Excellent chemical resistance to a wide variety of lab reagents
  • More transparent than polypropylene but with similar chemical resistance
  • Ideal for preparation and storage of culture media and titrations
  • All sizes have easy-to-read graduations
  • Autoclavable/Graduated

Includes:Polypropylene screw closures

TRUSTED_SUSTAINABILITY

Specifications

Capacity (English) 16 oz.
Description Flask Erlenmeyer Screw Closure PMP 500mL
Capacity (Metric) 500 mL
Material PMP
No. per Case 12
No. per Pack 4
Autoclavable Autoclavable
Disposable Reusable
Graduated Yes
Mouth Style Narrow
Sterility Non-sterile
Bottom Shape Plain
Closure Type Screw
Closure Size 43 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.