Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Polypropylene, End Caps, tri
GREENER_CHOICE

Catalog No. 12565976 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-565-976 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-565-976 Supplier Thermo Scientific™ Supplier No. 26650300
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For Use With: Nalgene Sanitary Carboys

Securely close off 3/4 in. or 3 in. sanitary ports on carboys with Thermo Scientific™ Nalgene™ Polypropylene End Caps. These are reliable, repeatable seals when used in combination with a silicone gasket and true union clamp. A groove on underside of end cap accepts standard sanitary gaskets for sealing the systems.

  • Seals sanitary ports on Nalgene Sanitary Carboys (No. 2630 and 2640)
  • USP Class VI
  • Autoclavable
TRUSTED_SUSTAINABILITY

Specifications

Closure Size 76 mm
Description 3 in. Tri (76mm)
For Use With (Equipment) Nalgene Sanitary Carboys
Material Polypropylene
Product Type Cap
Quantity Each
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.