Learn More
Thermo Scientific™ Nalgene™ Polypropylene, Replacement Coupling Inserts, elbow coupling, 1/4 inch
GREENER_CHOICE
Catalog No. 0292317
Change view
Click to view available options
Quantity:
Case of 6
Catalog No. Quantity
02-923-17 Case of 6
1 options
Catalog No. 02-923-17 Supplier Thermo Scientific™ Supplier No. 21590011
Add to Cart
Add to Cart
For Use With: Nalgene Quick Filling/Venting Closures
Promote faster installation using Thermo Scientific™ Nalgene™ Polypropylene Replacement Coupling Inserts for Quick/Filling Venting Closures, which support one-handed connection and disconnection when applied to mating coupling bodies. Male, barbed quick-disconnect replacement coupling inserts for use with Nalgene Quick Filling/Venting Closures.
  • For use with Nalgene Quick Filling/Venting Closures
  • Promotes fewer leakpoints and short assemblies
TRUSTED_SUSTAINABILITY
Specifications
Description 90° Elbow coupling insert
Diameter (English) Inner Tubing 1/4 in.
Diameter (Metric) Inner Tubing 6.35 mm
For Use With (Equipment) Nalgene Quick Filling/Venting Closures
Material Polypropylene
No. per Case 6/Cs.
No. per Pack 6 Pk.
Product Type Insert
Quantity Case of 6