Learn More
Thermo Scientific™ Nalgene™ Polypropylene, Replacement Coupling Inserts, elbow coupling, 3/8 inch
GREENER_CHOICE
Catalog No. 0292318 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
Case of 6
Catalog No. Quantity
02-923-18 Case of 6
1 options
Catalog No. 02-923-18 Supplier Thermo Scientific™ Supplier No. 21590012
Add to Cart
Add to Cart
For Use With: Nalgene Quick Filling/Venting Closures
Promote faster installation using Thermo Scientific™ Nalgene™ Polypropylene Replacement Coupling Inserts for Quick/Filling Venting Closures, which support one-handed connection and disconnection when applied to mating coupling bodies. Male, barbed quick-disconnect replacement coupling inserts for use with Nalgene Quick Filling/Venting Closures.
  • For use with Nalgene Quick Filling/Venting Closures
  • Promotes fewer leakpoints and short assemblies
TRUSTED_SUSTAINABILITY
Specifications
Description 90° Elbow coupling insert
Diameter (English) Inner Tubing 3/8 in.
Diameter (Metric) Inner Tubing 9.53 mm
For Use With (Equipment) Nalgene Quick Filling/Venting Closures
Material Polypropylene
No. per Case 6/Cs.
No. per Pack 6 Pk.
Product Type Insert
Quantity Case of 6