Learn More
Thermo Scientific™ Nalgene™ Polypropylene, Replacement Coupling Inserts, straight-through coupling, 1/4 inch
Shop All Thermo Scientific Products
Description
Promote faster installation using Thermo Scientific™ Nalgene™ Polypropylene Replacement Coupling Inserts for Quick/Filling Venting Closures, which support one-handed connection and disconnection when applied to mating coupling bodies. Male, barbed quick-disconnect replacement coupling inserts for use with Nalgene Quick Filling/Venting Closures.
- For use with Nalgene Quick Filling/Venting Closures
- Promotes fewer leakpoints and short assemblies
Specifications
Specifications
| Description | Straight-through coupling insert |
| Diameter (English) Inner Tubing | 1/4 in. |
| Diameter (Metric) Inner Tubing | 6.35 mm |
| For Use With (Equipment) | Nalgene Quick Filling/Venting Closures |
| Material | Polypropylene |
| No. per Case | 6/Cs. |
| No. per Pack | 6 Pk. |
| Product Type | Insert |