Learn More
Thermo Scientific™ Nalgene™ Polypropylene, Replacement Coupling Inserts, straight-through coupling, 3/8 inch
GSA_VAGREENER_CHOICE
Catalog No. 0292316 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
Case of 6
Catalog No. Quantity
02-923-16 Case of 6
1 options
Catalog No. 02-923-16 Supplier Thermo Scientific™ Supplier No. 21590002
Add to Cart
Add to Cart
For Use With: Nalgene Quick Filling/Venting Closures
Promote faster installation using Thermo Scientific™ Nalgene™ Polypropylene Replacement Coupling Inserts for Quick/Filling Venting Closures, which support one-handed connection and disconnection when applied to mating coupling bodies. Male, barbed quick-disconnect replacement coupling inserts for use with Nalgene Quick Filling/Venting Closures.
  • For use with Nalgene Quick Filling/Venting Closures
  • Promotes fewer leakpoints and short assemblies
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Description Straight-through coupling insert
Diameter (English) Inner Tubing 3/8 in.
Diameter (Metric) Inner Tubing 9.53 mm
For Use With (Equipment) Nalgene Quick Filling/Venting Closures
Material Polypropylene
No. per Case 6/Cs.
No. per Pack 6 Pk.
Product Type Insert
Quantity Case of 6