Learn More
Thermo Scientific™ Nalgene™ PPCO Centrifuge Bottles with Sealing Closure PROMO
Description
- Bottle includes polypropylene sealing cap assembly with silicone gasket for added leakproof assurance at maximum speeds or when spinning hazardous materials
- For proper performance, bottles must be filled to 80% of total capacity
- Requires use of the Nalgene centrifuge bottle adapter (DS3125-0250) when spinning 250 mL bottle in a Sorvall GSA rotor
Specifications
Specifications
| Capacity (Metric) | 450 mL |
| Capacity (English) | 16.9 oz. |
| Color | Translucent |
| Material | PPCO |
| Length (Metric) | 160 mm |
| No. per Pack | 4 Tubes/Pk. |
| Closure Included | Yes |
| Graduated | No |
| Max. Force | 13,700 x g |
| Shape | Round |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.