Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Rapid-Flow™ Sterile Disposable Bottle Top Filters with PES, CN, SFCA or Nylon Membranes
GSA_VAGREENER_CHOICE

Catalog No. 0974111
Change view
Click to view available options
Volume (Metric):
150 mL
500 mL
1000 mL
Material (Membrane):
PES
SFCA
Pore Size:
0.2 μm
0.45 μm
Fits Bottle Neck Size (Metric):
33mm
45mm
20 product options available for selection
Product selection table with 20 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Material (Membrane) Pore Size Fits Bottle Neck Size (Metric)
09-741-11 1000 mL PES 0.2 μm 45mm
09-741-08 150 mL PES 0.2 μm 33mm
09-741-09 150 mL PES 0.2 μm 45mm
09-740-62C 150 mL PES 0.45 μm 33mm
09-740-62D 150 mL PES 0.45 μm 45mm
09-741-06 500 mL PES 0.2 μm 33mm
09-741-07 500 mL PES 0.2 μm 45mm
09-740-64A 500 mL PES 0.45 μm 33mm
09-740-64B 500 mL PES 0.45 μm 45mm
09-741-10 1000 mL PES 0.2 μm 33mm
09-740-22D 150 mL SFCA 0.2 μm 33mm
09-740-22F 150 mL SFCA 0.2 μm 45mm
09-740-22E 150 mL SFCA 0.45 μm 33mm
09-740-22G 150 mL SFCA 0.45 μm 45mm
09-740-22H 500 mL SFCA 0.2 μm 33mm
09-740-22K 500 mL SFCA 0.2 μm 45mm
09-740-22J 500 mL SFCA 0.45 μm 33mm
09-740-22L 500 mL SFCA 0.45 μm 45mm
09-740-47A 1000 mL SFCA 0.2 μm 33mm
09-740-47C 1000 mL SFCA 0.2 μm 45mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
20 missing translation for 'options'
CAUTION: Use only sterile bottles designed for use with vacuum. Always use safety shield during vacuum procedures.
Catalog No. 09-741-11 missing translation for 'mfr' Thermo Scientific™ Supplier No. 5974520
Add to Cart
Add to Cart
CAUTION: Use only sterile bottles designed for use with vacuum. Always use safety shield during vacuum procedures.

Achieve fast flow rates and high throughput for increased filtration efficiency with Nalgene Rapid-Flow sterile disposable bottle top filters.

Increase filtration efficiency. Thermo Scientific™ Nalgene™ Rapid-Flow™ single use bottle tops feature the exclusive Rapid-Flow support column design, providing fast flow rates and high throughput. The last line of defense against cell culture contamination—Nalgene Rapid-Flow bottle tops are the best choice for the filtration of cell culture media, serum, additives and buffers.

  • Available in PES and SFCA membranes. Devices are color coded according to membrane.
  • Devices feature the fastest flow rate and lowest clogging of comparable alternatives
  • 0.1um PES available for mycoplasma protection
  • 0.2um PES devices are stem cell and mouse embryonic assay (MEA) tested
  • Non-pyrogenic and endotoxin free - Rapid-Flow filters meet the most rigorous testing standards for any vacuum filtration unit
  • Available with Fit for Purpose Sterile documentation

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Disposable Single-use
Filter Type Membrane Filter
Material Polystyrene
Material (Membrane) PES
Pore Size 0.2 μm
Sterility Sterile
Fits Bottle Neck Size (Metric) 45mm
No. per Case 12
No. per Pack 12
Type Bottle Top Filter
Volume (Metric) 1000 mL
Diameter (Metric) 90 mm
Diameter (Metric) Membrane 90 mm
Fit for Purpose Sterile
Show More Show Less
Can I remove the filter membranes from the Nalgene Rapid-Flow Sterile Disposable Bottle Top Filters with PES, CN, SFCA or Nylon Membranes?

No, the filter membranes for the bottle top filters are molded in and cannot be removed.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.