Learn More
- Home
- Products
- Filters and Filtration
- Filter Units
- Thermo Scientific™ Nalgene™ Rapid-Flow™ Sterile Disposable Filter Units with PES, CN, SFCA or Nylon Membranes
Thermo Scientific™ Nalgene™ Rapid-Flow™ Sterile Disposable Filter Units with PES, CN, SFCA or Nylon Membranes

Catalog No. 0974028D


| Catalog No. | Pore Size | Material (Membrane) | Diameter (Metric) Membrane | Volume (Metric) Receiver |
|---|---|---|---|---|
| 09-740-28D | 0.45 μm | SFCA | 75 mm | 500 mL |
| 09-741-88 | 0.2 μm | PES | 50 mm | 50 mL |
| 09-740-65B | 0.45 μm | PES | 50 mm | 115 mL |
| 09-741-200 | 0.1 μm | PES | 50 mm | 150 mL |
| 09-741-00 | 0.2 μm | PES | 50 mm | 115 mL |
| 09-741-01 | 0.2 μm | PES | 50 mm | 150 mL |
| 09-740-63A | 0.45 μm | PES | 50 mm | 150 mL |
| 09-741-201 | 0.1 μm | PES | 75 mm | 250 mL |
| 09-741-04 | 0.2 μm | PES | 50 mm | 250 mL |
| 09-740-63D | 0.45 μm | PES | 50 mm | 250 mL |
| 09-741-202 | 0.1 μm | PES | 75 mm | 500 mL |
| 09-741-02 | 0.2 μm | PES | 75 mm | 500 mL |
| 09-741-05 | 0.2 μm | PES | 90 mm | 500 mL |
| 09-740-63B | 0.45 μm | PES | 75 mm | 500 mL |
| 09-740-63E | 0.45 μm | PES | 90 mm | 500 mL |
| 09-741-203 | 0.1 μm | PES | 90 mm | 1000 mL |
| 09-741-03 | 0.2 μm | PES | 90 mm | 1000 mL |
| 09-740-63C | 0.45 μm | PES | 90 mm | 1000 mL |
| 09-740-36K | 0.2 μm | Cellulose Nitrate | 50 mm | 115 mL |
| 09-740-36L | 0.45 μm | Cellulose Nitrate | 50 mm | 115 mL |
| 09-740-1A | 0.2 μm | Cellulose Nitrate | 50 mm | 150 mL |
| 09-740-1B | 0.45 μm | Cellulose Nitrate | 50 mm | 150 mL |
| 09-740-1C | 0.8 μm | Cellulose Nitrate | 50 mm | 150 mL |
| 09-740-2A | 0.2 μm | Cellulose Nitrate | 50 mm | 250 mL |
| 09-740-2B | 0.45 μm | Cellulose Nitrate | 50 mm | 250 mL |
| 09-740-2C | 0.8 μm | Cellulose Nitrate | 50 mm | 250 mL |
| 09-740-25A | 0.2 μm | Cellulose Nitrate | 75 mm | 500 mL |
| 09-740-25B | 0.45 μm | Cellulose Nitrate | 75 mm | 500 mL |
| 09-740-25AA | 0.8 μm | Cellulose Nitrate | 75 mm | 500 mL |
| 09-740-3A | 0.2 μm | Cellulose Nitrate | 75 mm | 1000 mL |
| 09-740-3B | 0.45 μm | Cellulose Nitrate | 75 mm | 1000 mL |
| 09-740-3C | 0.8 μm | Cellulose Nitrate | 75 mm | 1000 mL |
| 09-740-36M | 0.2 μm | SFCA | 50 mm | 115 mL |
| 09-740-36N | 0.45 μm | SFCA | 50 mm | 115 mL |
| 09-740-28E | 0.2 μm | SFCA | 50 mm | 150 mL |
| 09-740-28F | 0.45 μm | SFCA | 50 mm | 150 mL |
| 09-740-39A | 0.2 μm | SFCA | 50 mm | 250 mL |
| 09-740-39B | 0.45 μm | SFCA | 50 mm | 250 mL |
| 09-740-28C | 0.2 μm | SFCA | 75 mm | 500 mL |
| 09-740-44A | 0.2 μm | SFCA | 90 mm | 500 mL |
| 09-740-44B | 0.45 μm | SFCA | 90 mm | 500 mL |
| 09-740-40A | 0.2 μm | SFCA | 75 mm | 500 mL |
| 09-740-43A | 0.2 μm | SFCA | 90 mm | 1000 mL |
| 09-740-40B | 0.45 μm | SFCA | 50 mm | 500 mL |
| 09-740-43B | 0.45 μm | SFCA | 90 mm | 1000 mL |
| 09-740-31F | 0.2 μm | Nylon | 50 mm | 150 mL |
| 09-740-31G | 0.45 μm | Nylon | 50 mm | 150 mL |
| 09-740-24A | 0.2 μm | Nylon | 50 mm | 250 mL |
| 09-740-24B | 0.45 μm | Nylon | 50 mm | 250 mL |
| 09-740-32 | 0.2 μm | Nylon | 75 mm | 500 mL |
| 09-740-45 | 0.2 μm | Nylon | 90 mm | 500 mL |
| 09-740-32B | 0.45 μm | Nylon | 75 mm | 500 mL |
| 09-740-26A | 0.2 μm | Nylon | 75 mm | 1000 mL |
| 09-740-46 | 0.2 μm | Nylon | 90 mm | 1000 mL |
| 09-740-26B | 0.45 μm | Nylon | 75 mm | 1000mL |
Description
Increase filtration efficiency. Thermo Scientific™ Nalgene™ Rapid-Flow™ Disposable Filter Units feature the exclusive Rapid-Flow support column design, providing fast flow rates and high throughput. The last line of defense against cell culture contamination—Nalgene Rapid Flow Filter Units and Bottle Tops are the best choice for the filtration of cell culture media, serum, additives and buffers. Some products on this page are ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions (see Full Specifications in the product table for item participation).
- Available in PES, CN, SFCA and Nylon membranes. Devices are color coded according to membrane.
- Devices feature the fastest flow rate and lowest clogging of comparable alternatives
- 0.1um PES available for mycoplasma protection
- 0.2um PES devices are stem cell and mouse embryonic assay (MEA) tested
- Non-pyrogenic and endotoxin free - Rapid-Flow filter units meet the most rigorous testing standards for any vacuum filtration unit.
- Available with Fit for Purpose Sterile documentation
- Some products on this page are ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions (see "Full Specifications" in the product table for item participation)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Frequently Asked Questions (FAQs)
We recommend using glass fiber prefilters for filtration of solutions with high viscosity or especially high particulate load. The glass fiber prefilters are available as an optional accessory and bundled with select Thermo Scientific Nalgene Rapid-Flow filter units. The prefilters enable a higher volume to be filtered before significant flow decay by trapping and preventing the larger particulate matter from entering the final filter membrane. Simply place one prefilter on top of the membrane bonded inside the unit, wet slightly to hold in place, and proceed with filtration as usual.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.