Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Rapid-Flow™ Sterile Disposable Filter Units with PES, CN, SFCA or Nylon Membranes
GSA_VAGREENER_CHOICE

Catalog No. 0974188
Change view
Click to view available options
Pore Size:
0.1 μm
0.2 μm
0.45 μm
0.8 μm
Material (Membrane):
Cellulose Nitrate
Nylon
PES
SFCA
Diameter (Metric) Membrane:
50 mm
75 mm
90 mm
Volume (Metric) Receiver:
50 mL
115 mL
150 mL
250 mL
500 mL
1000 mL
1000mL
55 product options available for selection
Product selection table with 55 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size Material (Membrane) Diameter (Metric) Membrane Volume (Metric) Receiver
09-741-88 0.2 μm PES 50 mm 50 mL
09-740-65B 0.45 μm PES 50 mm 115 mL
09-741-200 0.1 μm PES 50 mm 150 mL
09-741-00 0.2 μm PES 50 mm 115 mL
09-741-01 0.2 μm PES 50 mm 150 mL
09-740-63A 0.45 μm PES 50 mm 150 mL
09-741-201 0.1 μm PES 75 mm 250 mL
09-741-04 0.2 μm PES 50 mm 250 mL
09-740-63D 0.45 μm PES 50 mm 250 mL
09-741-202 0.1 μm PES 75 mm 500 mL
09-741-02 0.2 μm PES 75 mm 500 mL
09-741-05 0.2 μm PES 90 mm 500 mL
09-740-63B 0.45 μm PES 75 mm 500 mL
09-740-63E 0.45 μm PES 90 mm 500 mL
09-741-203 0.1 μm PES 90 mm 1000 mL
09-741-03 0.2 μm PES 90 mm 1000 mL
09-740-63C 0.45 μm PES 90 mm 1000 mL
09-740-36K 0.2 μm Cellulose Nitrate 50 mm 115 mL
09-740-36L 0.45 μm Cellulose Nitrate 50 mm 115 mL
09-740-1A 0.2 μm Cellulose Nitrate 50 mm 150 mL
09-740-1B 0.45 μm Cellulose Nitrate 50 mm 150 mL
09-740-1C 0.8 μm Cellulose Nitrate 50 mm 150 mL
09-740-2A 0.2 μm Cellulose Nitrate 50 mm 250 mL
09-740-2B 0.45 μm Cellulose Nitrate 50 mm 250 mL
09-740-2C 0.8 μm Cellulose Nitrate 50 mm 250 mL
09-740-25A 0.2 μm Cellulose Nitrate 75 mm 500 mL
09-740-25B 0.45 μm Cellulose Nitrate 75 mm 500 mL
09-740-25AA 0.8 μm Cellulose Nitrate 75 mm 500 mL
09-740-3A 0.2 μm Cellulose Nitrate 75 mm 1000 mL
09-740-3B 0.45 μm Cellulose Nitrate 75 mm 1000 mL
09-740-3C 0.8 μm Cellulose Nitrate 75 mm 1000 mL
09-740-36M 0.2 μm SFCA 50 mm 115 mL
09-740-36N 0.45 μm SFCA 50 mm 115 mL
09-740-28E 0.2 μm SFCA 50 mm 150 mL
09-740-28F 0.45 μm SFCA 50 mm 150 mL
09-740-39A 0.2 μm SFCA 50 mm 250 mL
09-740-39B 0.45 μm SFCA 50 mm 250 mL
09-740-28C 0.2 μm SFCA 75 mm 500 mL
09-740-44A 0.2 μm SFCA 90 mm 500 mL
09-740-28D 0.45 μm SFCA 75 mm 500 mL
09-740-44B 0.45 μm SFCA 90 mm 500 mL
09-740-40A 0.2 μm SFCA 75 mm 500 mL
09-740-43A 0.2 μm SFCA 90 mm 1000 mL
09-740-40B 0.45 μm SFCA 50 mm 500 mL
09-740-43B 0.45 μm SFCA 90 mm 1000 mL
09-740-31F 0.2 μm Nylon 50 mm 150 mL
09-740-31G 0.45 μm Nylon 50 mm 150 mL
09-740-24A 0.2 μm Nylon 50 mm 250 mL
09-740-24B 0.45 μm Nylon 50 mm 250 mL
09-740-32 0.2 μm Nylon 75 mm 500 mL
09-740-45 0.2 μm Nylon 90 mm 500 mL
09-740-32B 0.45 μm Nylon 75 mm 500 mL
09-740-26A 0.2 μm Nylon 75 mm 1000 mL
09-740-46 0.2 μm Nylon 90 mm 1000 mL
09-740-26B 0.45 μm Nylon 75 mm 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 55 options available.
55 options
Catalog No. 09-741-88 Supplier Thermo Scientific™ Supplier No. 5640020
Add to Cart
Edge
Add to Cart

Increase filtration efficiency. PES membranes and exclusive Rapid-Flow design provide fast flow rates and high throughput while defending against cell culture contamination.

Increase filtration efficiency. Thermo Scientific™ Nalgene™ Rapid-Flow™ Disposable Filter Units feature the exclusive Rapid-Flow support column design, providing fast flow rates and high throughput. The last line of defense against cell culture contamination—Nalgene Rapid Flow Filter Units and Bottle Tops are the best choice for the filtration of cell culture media, serum, additives and buffers. Some products on this page are ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions (see Full Specifications in the product table for item participation).

  • Available in PES, CN, SFCA and Nylon membranes. Devices are color coded according to membrane.
  • Devices feature the fastest flow rate and lowest clogging of comparable alternatives
  • 0.1um PES available for mycoplasma protection
  • 0.2um PES devices are stem cell and mouse embryonic assay (MEA) tested
  • Non-pyrogenic and endotoxin free - Rapid-Flow filter units meet the most rigorous testing standards for any vacuum filtration unit.
  • Available with Fit for Purpose Sterile documentation
  • Some products on this page are ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions (see "Full Specifications" in the product table for item participation)

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Disposable Single-use
Material (Membrane) PES
Pore Size 0.2 μm
Sterility Sterile
No. per Case 12
No. per Pack 12
Type Filter Unit
Volume (Metric) Receiver 50 mL
Volume (Metric) Upper 150 mL
Diameter (Metric) Membrane 50 mm
Fit for Purpose Sterile
Show More Show Less
What is the maximum volume that I can run through the Nalgene Rapid-Flow Sterile Disposable Filter Units with PES, CN, SFCA or Nylon Membranes?

The filters included with Nalgene Rapid-Flow Sterile Single Use Vacuum Filter Units are not specifically rated for a maximum volume that may be filtered. In many cases it depends on the specific application of the filter unit and would require testing beyond the volume of the receiver. Signs of the filter clogging, such as a decrease in filtration rate, would likely indicate the need for replacement.

I am using a Nalgene Rapid-Flow Sterile Disposbable Filter Unit and my solution is taking a long time to filter. Do you have any recommendations to improve filtering?

We recommend using glass fiber prefilters for filtration of solutions with high viscosity or especially high particulate load. The glass fiber prefilters are available as an optional accessory and bundled with select Thermo Scientific Nalgene Rapid-Flow filter units. The prefilters enable a higher volume to be filtered before significant flow decay by trapping and preventing the larger particulate matter from entering the final filter membrane. Simply place one prefilter on top of the membrane bonded inside the unit, wet slightly to hold in place, and proceed with filtration as usual.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.