Learn More
Thermo Scientific™ Nalgene™ Rapid-Flow™ Sterile Filter Storage Bottles



Description
Save time and minimize contamination with gamma-sterilized Thermo Scientific™ Nalgene™ Rapid-Flow™ Filter Storage Bottles. Use with Nalgene Bottle-Top filters or as replacement/extra receptacles for filter units. Excellent as single-use storage containers.
- Headspace allows for extra serum or other additives to be added to the filtrate
- Leakproof† screw caps to eliminate pH shift in bottle for longer life of stored media
- Sterilized by gamma radiation and individually bagged for a 5-year sterile shelf life
- Non-pyrogenic and non-cytotoxic
- Available with Fit for Purpose Sterile documentation
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Cap Material | Virgin HDPE |
| Closure Color | White |
| Material | Virgin Polystyrene |
| Sterility | Sterile |
| Model | Wide |
| Mouth | Wide |
| No. per Case | 12 |
| No. per Pack | 12 |
| Volume (Metric) | 1 L |
| Fit for Purpose | Sterile |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.