Learn More
Beckman Coulter Nalgene Sample Cup Tube Adaptor, 5 mL
Adapter allows the proper alignment of a 5 mL Nalgene Sample Cup (6250-0005) on the Accuvette stage. Sample cups are crucial for precious samples, or when there is limited sample material available.
Supplier:  Beckman Coulter A93166
Catalog No. 15-655-131
Only null left
Add to Cart
Description
Description
  • Designed for use with 5 mL Nalgene Sample Cups (Nalgene 6250-0005)
  • Fits on top of the platform in the Accuvette holder
  • Ideal for precious samples, or when minimal sample material is available
Specifications
Specifications
Tube Adaptor
1 Ea.
Nalgene Sample Cup, 5 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.