Learn More
Thermo Scientific™ Nalgene™ Centrifuge Tube Sealing Cap Assemblies
Shop All Thermo Scientific Products

Description
Process Thermo Scientific™ Nalgene™ FEP Oakridge 30mL and 50mL centrifuge tubes at their highest rated speed level (10,000 x g and over) without leakage, even when full. Thermo Scientific™ Nalgene™ Sealing Cap Assemblies include a Viton™ fluoroelastomer o-ring. Also recommended when spinning hazardous materials compatible with centrifuge tube material.
Features:
- Top fits securely for use at maximum speeds without leakage
- Autoclavable/biohazard/leakproof†
Includes:
- Polypropylene screw closure
- Tefzel™ ETFE plug
- Viton fluoroelastomer o-ring
Specifications
Specifications
| Closure Size | 24 mm |
| Product Type | Centrifuge Tube Cap |
| Autoclavable | Autoclavable |
| Description | Sealing cap assembly |
| Material | Polypropylene, ETFE, Fluoroelastomer |
| No. per Pack | 2 |
| No. per Case | 2 |
| Product Line | Nalgene |
| For Use With (Equipment) | Closure size 24 |
Frequently Asked Questions (FAQs)
Replacement sealing-cap assemblies are available for selected sizes. We recommend Nalgene Centrifuge Tube Sealing Cap Assemblies (Cat. No. DS3131-0020) for 30 mL tubes and Nalgene Centrifuge Tube Sealing Cap Assemblies (Cat. No. DS3131-0024) for 50 mL tubes.
The actual brim capacity of the Nalgene Oak Ridge High-Speed PPCO Centrifuge Tubes with Sealing Cap is 42 mL, but it is designed to fit a standard rotor.
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.