Learn More
Thermo Scientific™ Nalgene™ Sealing Caps for Oak Ridge Centrifuge Tubes; PP screw closure, silicone gasket
Shop All Thermo Scientific Products
Description
- Polypropylene closures with silicone gasket
Before autoclaving, set closure on top of the container without engaging the threads.
Specifications
Specifications
| Autoclavable | Autoclavable |
| Material | Silicone |
| Tip Material | Silicone |
| For Use With (Equipment) | Replacement O-ring (for Mfr. No. DS3132-0058) |