Learn More
Thermo Scientific™ Nalgene™ Sealing Caps for Oak Ridge Centrifuge Tubes

Shop All Thermo Scientific Products


Description
Ensure leakproof† service of Thermo Scientific™ Nalgene™ centrifuge bottles at their recommended maximum rated speeds by using Nalgene Sealing Caps for Oak Ridge Centrifuge Tubes. They are also recommended for use when spinning hazardous materials.
Compatibility:
- DS3132-0058: for use with Nalgene Centrifuge Bottles 3140-0250 and 3140-0250
- DS3132-0063: ror use with Nalgene Centrifuge Bottles 3140-0500, 3141-0500, 3120-1000, 3120-1010, 3122-1000 and 3122-1010
- DS3132-0020: for use with Nalgene Oakridge tubes, 28ml and 30ml volumes
- DS3132-0024: for use with Nalgene Oakridge tubes, 35ml and 50ml volumes
Note:
Before autoclaving, set closure on top of the container without engaging the threads.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Closure Size | 24 mm |
| Product Type | Centrifuge Tube Cap |
| Autoclavable | Autoclavable |
| Description | Sealing Caps for Oak Ridge Centrifuge Tubes |
| Material | Polypropylene, Rubber |
| No. per Pack | 2 |
| No. per Case | 2 |
| Product Line | Nalgene |
| For Use With (Equipment) | 50 mL Tubes |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.