Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Single-Use PETG Erlenmeyer Flasks with Baffled Bottom: Sterile
GREENER_CHOICE

Catalog No. 03600153
Change view
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
1000 mL
2000 mL
2800 mL
Closure Size:
70 mm
38-430 mm
45-430 mm
Closure Type:
Solid, non-vented
Vented
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Size Closure Type
03-600-153 2800 mL 70 mm Vented
10-041-23 125 mL 38-430 mm Solid, non-vented
10-346-8A 250 mL 38-430 mm Solid, non-vented
10-346-8B 500 mL 45-430 mm Solid, non-vented
10-041-24 1000 mL 45-430 mm Solid, non-vented
10-346-8C 2000 mL 45-430 mm Solid, non-vented
03-600-151 2800 mL 70 mm Solid, non-vented
10-529 125 mL 38-430 mm Vented
10-530 250 mL 38-430 mm Vented
10-531 500 mL 45-430 mm Vented
10-532 1000 mL 45-430 mm Vented
10-533 2000 mL 45-430 mm Vented
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Catalog No. 03-600-153 Supplier Thermo Scientific™ Supplier No. 41162800
Add to Cart
Add to Cart
This item is not returnable. View return policy

Save preparation time and avoid contamination risk with these single-use flasks, ideal for suspension of cell culture, media preparation, mixing, and storage.

Save preparation time and avoid contamination risk by using Thermo Scientific Nalgene Single-Use PETG Erlenmeyer Flasks with Baffled Bottom, ideal for suspension of cell culture, media preparation, mixing, and storage. Crystal-clear PETG is robust and break resistant, increasing personal safety. All sizes are available vented or non-vented.

Features

  • Sterile and single use to reduce contamination risk
  • BPA-free
  • Collapses when autoclaved to reduce bio hazardous waste
  • Graduated for quick volume assessment

Warranty: 90 days

TRUSTED_SUSTAINABILITY

Specifications

Capacity (English) 95 oz.
Capacity (Metric) 2800 mL
Material PETG
No. per Case 4
No. per Pack 1
Shape Fernbach
Surface Coating None
Green Features ACT Ecolabel (Silver)
Autoclavable Not Autoclavable
Disposable Single-use
Graduated Yes
Mouth Style Narrow
Sterility Sterile
Bottom Shape Baffled
Closure Type Vented
Closure Size 70 mm
Show More Show Less

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.