Learn More
Thermo Scientific™ Nalgene™ Stainless-Steel Forceps, bent tip
GSA_VAGREENER_CHOICE
Catalog No. 09742 Shop All Thermo Scientific Products
Change view
Click to view available options
Tip Style:
Bent, Round, Serrated
Catalog No. Tip Style
09-742 Bent, Round, Serrated
1 options
Catalog No. 09-742 Supplier Thermo Scientific™ Supplier No. DS03990001
  • $117.70
    $73.65 / Case of 1
    Save $44.05
    37% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Nalgene Stainless-Steel Forceps are available in bent or straight-tip styles. Bent-tip style is designed for easy removal and manipulation of filter membranes.
Designed for use in a wide range of applications, Thermo Scientific™ Nalgene™ filtration hardware and accessories can be used in conjunction with our various lines of filtration devices.
  • Stainless steel
  • Self-closing (bent tip)
  • Serrated gripping area
  • Autoclavable
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Material Stainless Steel
Description Stainless-Steel Forceps, bent tip
No. per Pack 1
Quantity Ea.
Style Self-closing Bent
Tip Style Bent, Round, Serrated
Product Type Forceps