Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Teflon™ Low Particulate Bottles
GSA_VAGREENER_CHOICE

Catalog No. 03313106
Change view
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
1 L
Closure Size:
24 mm
28 mm
38 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Size
03-313-106 500 mL 28 mm
03-313-104 125 mL 24 mm
03-313-105 250 mL 24 mm
03-313-107 1 L 38 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 03-313-106 Supplier Thermo Scientific™ Supplier No. 3816000016
Add to Cart
Add to Cart

Nalgene Teflon Low Particulate Bottles are certified clean to USP <788> standards, making them an ideal solution for biological and pharmaceutical manufacturing.

Sample and store active pharmaceutical ingredients and bulk intermediates optimally with Thermo Scientific™ Nalgene™ Low Particulate/Low Metals Bottles Made of Teflon™ FEP with Closure, certified to contain less than 20 particles per mL at 0.3 μm and greater. Metals certified to micrograms per liter (ppb) levels. Excellent for storing high purity chemicals, this product includes a certificate of quality that assures the product has been tested and accepted in accordance with specifications.

Features:

  • Certified to particulate and trace metals levels
  • Linerless Tefzel™ ETFE closures provide the ultimate in leakproof† protection without the use of liner that can wrinkle, cause leaks, or contaminate your reagents
  • Exhibits flexibility, impact resistance and excellent visibility of contents for a wide variety of uses in the lab
  • Excellent chemical resistance of almost any lab chemical
  • Excellent for storing high purity chemicals
  • Non-sterile/autoclavable
  • Translucent

Each bottle is double bagged under Class 10 laminar flow hoods inside a Class 100 clean room. Excellent for ICP-MS reagent and standard storage. Product includes a certificate of quality that assures the product has been tested and accepted in accordance with specifications.

Metals certified to microliter (ppb) levels: <0.20 Hg, <0.5 Be, <1.0 As/Cd/Pb, <2.0 Sb/Se, <5.0 Ag/Co/Cr/Cu/Mn/Tl/V, <10 Ba/Ni/Zn, <50 Mg, <75 Al, <100 Ca/Fe/K/Na

Warranty: 90 days

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 500 mL
Capacity (English) 16 oz.
Color Clear
Diameter (Metric) Inner Neck 20 mm
For Use With (Application) Storage, Transportation ​
Material Teflon FEP
No. per Case 4/Cs.
No. per Pack 1 Ea.
Sterility Non-sterile
Temperature Range (Metric) -100°C to 260°C
Closure Color White
Low Particulate USP <788> (Low particulate)
Neck Finish 28 mm
Certifications/Compliance Certified to particulate and trace metals levels. Metals certified to microliter (ppb) levels <0.20 Hg, <0.5 Be, <1.0 As/Cd/Pb, <2.0 Sb/Se, <5.0 Ag/Co/Cr/Cu/Mn/Tl/V, <10 Ba/Ni/Zn, <75 Al, <100 Ca/Fe/K/Na. Certified to contain <20 particles
Closure Included Yes
Graduated No
Mouth Narrow
Shape Round
Height (Metric) 166 mm
Product Type Low Particulate/Low Metals Bottle
Width (English) 2.86 in.
Height (English) 6.53 in.
Closure Liner Unlined
Closure Material Tefzel™ ETFE
Closure Size 28 mm
Diameter (Metric) 73 mm
Closure Type Screw Cap
Through the Closure Dispensing No
Autoclavable Autoclavable
Workflow Step Upstream process solution storage. Upstream Validation. Downstream process solution storage. Downstream Validation
Show More Show Less
Why do you recommend Teflon-coated bottles to store high-value contents such as, active pharmaceutical ingredients (API's), bulk intermediates, and critical reagents?

Teflon is ideal for storing the highest-value contents because it is resistant to most chemicals, temperatures, and processes, allowing for rigorous cleaning procedures such as those that require boiling nitric acid. Teflon also allows you to store contents down to cryogenic temperatures for products requiring ultra-low temperature (below -100 degrees C) storage.
We offer Nalgene Teflon bottles that provide excellent chemical and temperature resistance in a leak-proof design. They are manufactured using quality resins, with the added value of high-level support, making Nalgene Teflon bottles an ideal choice for your high-value contents.

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.