Learn More
Thermo Scientific™ Nalgene™ Tubing Y-Type Connectors
Description
Maximize liquid flow with non-tapered hose barbed Thermo Scientific™ Nalgene™ Tubing Y-Type Connectors, molded of polypropylene resin, free of animal derived components. Products are double-bagged to maintain cleanliness.
- Polypropylene construction
- Autoclavable
Specifications
Specifications
| Description | Y-Type Connector |
| Autoclavable | Autoclavable |
| Diameter (English) Inner | 1/2 in. |
| Diameter (Metric) Inner | 12.7 mm |
| Material | Polypropylene |
| For Use With | 1/2 in. Tubing |
| Packaging Quantity | 12/Pk., 48/Cs. |
| Product Type | Tubing Connector |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.