Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medisca Naloxone Hydrochloride USP/EP (Dihydrate)
SDP

Catalog No. 14186202 Shop All Medisca Products
Change view
Click to view available options
Quantity:
500 mg
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-186-202 500 mg
14-185-957 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-186-202 Supplier Medisca Supplier No. 076700
Add to Cart
Add to Cart

Naloxone Hydrochloride USP / EP is a white to slightly off-white powder. It is hygroscopic. Alternative name(s): naloxone hydrochloride dihydrate

Naloxone Hydrochloride USP / EP is a white to slightly off-white powder. It is hygroscopic.

Alternative name(s): naloxone hydrochloride dihydrate

Specifications

Chemical Name or Material Naloxone Hydrochloride USP/EP (Dihydrate)
Packaging Bottle, Amber Glass
CAS 51481-60-8
Type Drug Standard
Molecular Weight (g/mol) 399.8684
Quantity 500 mg
Percent Purity 98 to 100.5 %
Grade EP, USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.