Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NALP5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143281
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143281 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5143281 Supplier Invitrogen™ Supplier No. PA5143281
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.

NALP proteins include the apoptosis regulator APAF1 (apoptotic protease activating factor 1) and mammalian NOD-LRR proteins and are thought to be involved in inflammation and reproduction. NALP5, also known as MATER, is a maternal gene required for early embryonic development in mice. Increased NALP5 expression was observed in two neuronal injury models, and transient expression of recombinant NALP5 in neurons induced caspase-3 activation and apoptosis, suggesting that NALP5 also plays a role in caspase activation and apoptosis in injured neurons, and may thus represent a novel target for therapeutic treatment in neurodegenerative disorders.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NALP5
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene NLRP5
Gene Accession No. P59047
Gene Alias CLR19.8; MATER; Mater protein; Mater protein homolog; maternal antigen that embryos require; NACHT, leucine rich repeat and PYD containing 5; NACHT, LRR and PYD domains-containing protein 5; Nalp5; NLR family pyrin domain containing 5; NLR family, pyrin domain containing 5; NLRP5; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 5; Ooplasm-specific protein 1; OP1; PAN11; PYPAF8
Gene Symbols NLRP5
Host Species Goat
Immunogen Peptide with sequence C-DGSWHSFDEDDR.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126206
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.