Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NLRP6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560466
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560466 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560466 Supplier Invitrogen™ Supplier No. PA560466
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GEEPNYPLEL LYCLYETQED AFVRQALCRF PELALQRVRF CRMDVAVLSY CVRCCPAGQA LRLISCRLVA AQEKKKKSLG KRLQASLGGG SSSQGTTKQL PASLLHPLFQ Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 65%.

NALP proteins include the apoptosis regulator APAF1 (apoptotic protease activating factor 1) and mammalian NOD-LRR proteins and are thought to be involved in inflammation and reproduction. NALP6, also known as PYPAF5, is highly expressed in granulocytes and T cells and co-localizes with the PYRIN-CARD protein ASC. Co-expression of NALP6 with ASC results in a synergistic activation of NF-kappa-B and activated caspase-1-dependent cytokine processing, suggesting that NALP6 functions in immune cells to coordinate the transduction of pro-inflammatory signals.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NLRP6
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NLRP6
Gene Accession No. P59044
Gene Alias AI504961; angiotensin II/vasopressin receptor; angiotensin/vasopressin receptor; AVR; CLR11.4; dual angiotensin II/vasopressin receptor; NACHT, leucine rich repeat and PYD containing 6; NACHT, LRR and PYD domains-containing protein 6; NACHT/LRR/pyrin domain-containing protein 6; NALP6; Navr; Navr/Avr; NLR family pyrin domain containing 6; NLR family, pyrin domain containing 6; Nlrp6; Non-angiotensin-vasopressin receptor; Non-AVR; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 6; PAN3; Pypaf5; PYRIN-containing APAF1-like protein 5; PYRIN-containing APAF1-like protein 5-like
Gene Symbols NLRP6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NALP6. Recombinant protein control fragment (Product #RP-97487).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171389
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.