Learn More
Description
Immunogen sequence: CRLTEASCKD LAAVLVVSKK LTHLCLAKNP IGDTGVKFLC EGLSYPDCKL QTLVLQQCSI TKLGCRYLSE ALQEACSLTN LDLSINQIAR GLWILCQALE NPNCNLKHLR LWSCSLMPFY CQHLGSALLS NQKLETLDLG QNHLWKSGII KLFGVLRQRT GSLKILRLKT YETNLEIKKL LEEVKEKNPK LTIDCNASGA T; Positive Samples: HeLa, U-87MG, 293T, Jurkat, Mouse brain.
Specifications
Specifications
| Antigen | NALP7 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 2.24 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | NLRP7 |
| Gene Accession No. | Q8WX94 |
| Gene Alias | CLR19.4; HYDM; NACHT, leucine rich repeat and PYD containing 7; NACHT, LRR and PYD containing protein 7; NACHT, LRR and PYD domains-containing protein 7; NALP7; NLR family pyrin domain containing 7; NLR family, pyrin domain containing 7; NLRP7; NOD12; Nucleotide-binding oligomerization domain protein 12; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 7; PAN7; PYPAF3; PYRIN-containing APAF1-like protein 3 |
| Gene Symbols | NLRP7 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
